Skip to Content
Merck
All Photos(1)

Documents

NIST76A

Burnt refractory

NIST® SRM® 76a, Al203-40%

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

powder

packaging

pkg of 75 g

manufacturer/tradename

NIST®

Looking for similar products? Visit Product Comparison Guide

General description

Standard Reference Materials (SRMs) NIST76a, NIST77a and NIST78a are in the form of fine powder (<0.15 mm) and are intended for use in checking chemical methods of analysis and in calibration with instrumental methods of analysis. For more information, please refer to the related SDS and COA.

SRM 76A_cert

SRM 76A_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Aluminium oxide (Al2O3), Calcium oxide (CaO), Iron (III) Oxide (Fe2O3), Lithium oxide (Li2O), Magnesium oxide (MgO), Phosphorus pentoxide (P2O5), Potassium oxide (K2O), Silicon dioxide (SiO2), Sodium oxide (Na2O), Strontium oxide (SrO), Titanium dioxide (TiO2), Calcium oxide (CaO)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - STOT RE 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service