grade
certified reference material
Quality Level
packaging
pkg of 4 wafers
manufacturer/tradename
NIST®
application(s)
glass & ceramic
format
matrix material
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Other Notes
Antimony (Sb), Arsenic (As), Barium (Ba), Cadmium (Cd), Chromium (Cr), Iron (Fe), Lead (Pb), Manganese (Mn), Nickel (Ni), Rubidium (Rb), Selenium (Se), Silver (Ag), Strontium (Sr), Thorium (Th), Uranium (U)
Reference/Informational value is provided for the following analytes.
Cobalt (Co), Copper (Cu), Thallium (Tl), Titanium (Ti), Boron (B), Cerium (Ce), Dysprosium (Dy), Erbium (Er), Europium (Eu), Gadolinium (Gd), Gold (Au), Lanthanum (La), Lithium (Li), Neodymium (Nd), Potassium (K), Samarium (Sm), Ytterbium (Yb)
See certificate for values and more details at nist.gov/SRM.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 1B Inhalation
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service