Skip to Content
Merck
All Photos(1)

Key Documents

NIST2806B

Medium test dust (MTD) in hydraulic fluid

NIST® SRM® 2806b

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

packaging

pkg of 2 x 400 mL

manufacturer/tradename

NIST®

application(s)

pharmaceutical (small molecule)

format

matrix material

General description

This Standard Reference Material (SRM) is intended for use in the calibration of instrumental response to medium test dust suspended in hydraulic fluid. A unit of SRM 2806b consists of two bottles containing polydisperse, irregularly shaped mineral dust suspended in approximately 400 mL of hydraulic fluid each. For more information, please refer to the COA and SDS.

SRM 2806B_cert SRM 2806B _SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

168.8 °F

Flash Point(C)

76 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service