Skip to Content
Merck
All Photos(2)

Key Documents

N9405

Millipore

Nutrient agar

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
R$1,859.00

R$1,859.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 G
R$1,859.00

About This Item

UNSPSC Code:
41171605
NACRES:
NA.85

R$1,859.00


Check Cart for Availability

Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

Agar, 15.00 g/L
Beef extract, 3.00 g/L
Peptic digest of animal tissue, 5.00 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

pH

6.8±0.2 (25 °C)

application(s)

agriculture
bioburden testing
clinical testing
environmental
food and beverages
water monitoring

microbiology

suitability

nonselective for Enterococcus spp.
nonselective for Escherichia coli
nonselective for Salmonella spp.
nonselective for Shigella spp.
nonselective for Staphylococcus spp.
nonselective for bacteria (General Media)
nonselective for coliforms

Application

Used for the cultivation of bacteria and for the enumeration of organisms in water, sewage and feces.

Preparation Note

Suspend 23 g of Nutrient Agar pH 6.8 in 1000 ml of distilled water. Boil to dissolve the medium completely. Sterilize by autoclaving at 15 lbs. pressure (121°C) for 15 minutes. If desired, the medium can be enriched with 5-10% v/v sterile defibrinated blood.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Michele E Barbour et al.
International journal of nanomedicine, 8, 3507-3519 (2013-10-05)
Chlorhexidine (CHX) is an antimicrobial agent that is efficacious against gram-negative and -positive bacteria and yeasts. Its mechanism of action is based on cell membrane disruption and, as such, it does not promote the development of bacterial resistance, which is
Greenberg, A.E., et al.
Standard Methods for Examination of Water and Wastewater (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service