Skip to Content
Merck
All Photos(1)

Key Documents

N2B

Low Temperature Viscosity Standard

ASTM D 2162 traceable

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
R$1,363.00

R$1,363.00


Estimated to ship onMay 17, 2025


Request a Bulk Order

Select a Size

Change View
500 ML
R$1,363.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

R$1,363.00


Estimated to ship onMay 17, 2025


Request a Bulk Order

grade

certified reference material

quality

ASTM D 2162 traceable

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon Low Temperature Viscosity Standards can be found on the Paragon Scientific webpage
This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service