Skip to Content
Merck
All Photos(3)

Key Documents

M2047

Sigma-Aldrich

Methyl salicylate

BioXtra

Synonym(s):

2-Hydroxybenzoic acid methyl ester, Methyl 2-hydroxybenzoate, Oil of wintergreen, Wintergreen oil

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CO2CH3
CAS Number:
Molecular Weight:
152.15
Beilstein:
971516
EC Number:
MDL number:
UNSPSC Code:
41116100
PubChem Substance ID:
NACRES:
NA.25

vapor density

5.26 (vs air)

Quality Level

vapor pressure

1 mmHg ( 54 °C)

product line

BioXtra

form

liquid

autoignition temp.

847 °F

impurities

≤0.0005% Phosphorus (P)

ign. residue

≤0.1%

refractive index

n20/D 1.536 (lit.)

bp

222 °C (lit.)

mp

−8-−7 °C (lit.)

density

1.174 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

COC(=O)c1ccccc1O

InChI

1S/C8H8O3/c1-11-8(10)6-4-2-3-5-7(6)9/h2-5,9H,1H3

InChI key

OSWPMRLSEDHDFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Repr. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Agata Bartyzel et al.
Analytica chimica acta, 707(1-2), 204-209 (2011-10-27)
N,N'-bis-(α-methylsalicylidene)-2,2-dimethyl-1,3-propanediimine (SBTD) modified silica gel was prepared and used as sorbent for solid phase extraction of Cr(III) ions from aqueous solution. This sorbent showed a high sorption affinity for Cr(III) while recovery of Cr(VI) was very low. The analyte ion
Nadir Erbilgin et al.
Tree physiology, 32(8), 946-957 (2012-06-05)
Coniferous trees have both constitutive and inducible defences that deter or kill herbivores and pathogens. We investigated constitutive and induced monoterpene responses of jack pine (Pinus banksiana Lamb.) to a number of damage types: a fungal associate of the mountain
Michiel van Wijk et al.
PloS one, 6(7), e21742-e21742 (2011-07-19)
Insect herbivory induces plant odors that attract herbivores' natural enemies. Assuming this attraction emerges from individual compounds, genetic control over odor emission of crops may provide a rationale for manipulating the distribution of predators used for pest control. However, studies
Youli Yao et al.
The Plant cell, 23(10), 3842-3852 (2011-10-27)
We have previously shown that local exposure of plants to stress results in a systemic increase in genome instability. Here, we show that UV-C-irradiated plants produce a volatile signal that triggers an increase in genome instability in neighboring nonirradiated Arabidopsis
Xinhua Zhang et al.
Journal of agricultural and food chemistry, 59(17), 9351-9357 (2011-07-28)
The effects of methyl salicylate (MeSA) on chilling injury (CI) and gene expression levels, enzyme activities, and metabolites related to arginine catabolism in cherry tomato fruit were investigated. Freshly harvested fruits were treated with 0.05 mM MeSA vapor at 20

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service