IRMMIFCC467
Hemoglobin HbA0 (non-glycated)
IRMM®, certified reference material
Synonym(s):
Haemoglobin isolated from whole blood
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material
Agency
IRMM®
manufacturer/tradename
JRC
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
−70°C
Analysis Note
For more information please see:
IRMMIFCC467
IRMMIFCC467
Legal Information
IRMM is a registered trademark of European Commission
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service