Skip to Content
Merck
All Photos(1)

Key Documents

FA10006P

Sigma-Aldrich

PIPETMAN® P1000L Micropipette Plastic eject

for use with BioControl® GDS, suitable for PCR, tip volume 100-1000 μL

Synonym(s):

PIPETMAN P1000L Micropipette Plastic eject, GDS-compatible micropipette, Micropipette for GDS system

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
R$2,791.00

R$2,791.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 EACH
R$2,791.00

About This Item

UNSPSC Code:
41121521
NACRES:
NB.02

R$2,791.00


Check Cart for Availability

Request more information

packaging

box of 1 unit

technique(s)

PCR: suitable

tip volume

100-1000 μL

compatibility

for use with BioControl® GDS

storage temp.

room temp

General description

PIPETMAN® P1000L is a versatile laboratory tool that can be used in conjunction with the Assurance GDS® (Genetic Detection System). Assurance GDS® is an automated nucleic acid amplification system for the detection of pathogens in food, ingredients, and environmental samples. The P1000L Micropipette plastic eject is an air-cushion pipette with outstanding precision and accuracy. It is extremely reliable and robust, allowing for long service life. The PIPETMAN® provides fast and ergonomic handling with reduced operating efforts and active aerosol reduction. The Micropipette plastic eject is used for accurate and precise liquid handling in the microliter range (up to 1000 µL). The PIPETMAN® P1000L features a durable plastic tip ejector that allows for quick and easy disposal of pipette tips after each use.

Application

PIPETMAN® P1000L Micropipette plastic eject is used to transfer PCR reagents with Assurance GDS® to complete sample preparation for the PCR detection of pathogenic organisms in foods, ingredients, and environmental samples.

Components

The Product Contains the following:
  • Single Micropipette.
  • Plastic Rejector.

Legal Information

Assurance GDS is a registered trademark of BioControl Systems, Inc.
BioControl is a registered trademark of BioControl Systems, Inc.
Pipetman is a registered trademark of Gilson S.A.S. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service