Skip to Content
Merck
All Photos(1)

Key Documents

F6270

Supelco

Fipexide hydrochloride

analytical standard

Synonym(s):

1-(2-[4-Chlorophenoxy]acetyl)-4-(3,4-methylenedioxybenzyl)piperazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21ClN2O4 · HCl
CAS Number:
Molecular Weight:
425.31
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

Cl.Clc1ccc(OCC(=O)N2CCN(CC2)Cc3ccc4OCOc4c3)cc1

InChI

1S/C20H21ClN2O4.ClH/c21-16-2-4-17(5-3-16)25-13-20(24)23-9-7-22(8-10-23)12-15-1-6-18-19(11-15)27-14-26-18;/h1-6,11H,7-10,12-14H2;1H

InChI key

MVOWQBJZZKOQNU-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Genkova-Papasova et al.
Acta physiologica et pharmacologica Bulgarica, 14(4), 36-41 (1988-01-01)
Experiments on albino rats were carried out to study the effects of the nootropic drugs piracetam, aniracetam, meclofenoxate and fipexide on the DA-beta-hydroxylase inhibitor diethyldithiocarbamate- and the alpha 2-adrenoceptor agonist clonidine-impaired cognitive functions. The changes in memory were studied by
[Fipexide: a new drug for the regulation of dopaminergic system at the macromolecular level].
C Missale et al.
Bollettino chimico farmaceutico, 122(2), 79-85 (1983-02-01)
F Durand et al.
Journal of hepatology, 15(1-2), 144-146 (1992-05-01)
Fipexide belongs to a new class of cognition activators and is noted for its lack of amphetamin-like side effects. We describe three patients who developed fulminant hepatic failure less than 2 months after beginning fipexide administration. The mean interval from
[Acute drug-induced hepatitis: a case of acute cytolysis after ingestion of fipexide].
F Mion et al.
Gastroenterologie clinique et biologique, 14(5), 513-514 (1990-01-01)
A Marino et al.
Pharmacological research, 22(2), 179-187 (1990-03-01)
Dopamine neurotransmission plays a role in learning, memory and related cognitive processes. We evaluated the effect of a nootropic drug, fipexide, a parachloro-phenossiacetic acid derivative, both on behavioural and biochemical parameters. The compound was able to cause an improved performance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service