Skip to Content
Merck
All Photos(1)

Key Documents

EH0030003764BC

Conductive Disposable Tips

BioControl, For use with Gemini, size 300 μL

Synonym(s):

Disposable Tips; Pipette Tips, Robotic pipette tips

Sign Into View Organizational & Contract Pricing

Select a Size

960 UNIT
R$1,080.00

R$1,080.00


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
960 UNIT
R$1,080.00

About This Item

UNSPSC Code:
41122409
NACRES:
NB.02
volume:
300 μL
packaging:
box of 960 tips
manufacturer/tradename:
BioControl Systems E-H0030-003-764*
Gemini®

R$1,080.00


Check Cart for Availability

Request a Bulk OrderRequest more information

packaging

box of 960 tips

manufacturer/tradename

BioControl Systems E-H0030-003-764*
Gemini®

technique(s)

ELISA: suitable
enzyme immunoassay: suitable

size

300 μL

volume

300 μL

application(s)

food and beverages
pathogen testing

compatibility

for use with Gemini (78000BC)

storage temp.

room temp

General description

The Conductive Disposable Tips are specially designed for use with automated liquid handling platforms. The tips can hold a volume of 300 μl and display tighter tolerances when compared to tips used for handheld pipettes. They enable an increase in pipetting performance due to straighter tips and lower percentage coefficient of variation (CV) of the tips. The Conductive Disposable Tips are packaged for easy use and are readily compatible with specific automated systems. In this case for use with Gemini®.

Application

The Conductive Disposable Tips used with Gemini® provide automation of ELISA and other diagnostic Enzyme Immunoassays (EIA).

Components

Conductivity Tips, 300 μL, 960

Legal Information

Gemini is a registered trademark of Stratec

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service