Skip to Content
Merck
All Photos(1)

Key Documents

E2205500

1,1′-Ethylidenebistryptophan

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1,1′-Ethylidenebis-(L-tryptophan), EBT

Sign Into View Organizational & Contract Pricing

Select a Size

2 EA
R$1,229.00

R$1,229.00


Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
2 EA
R$1,229.00

About This Item

Empirical Formula (Hill Notation):
C24H26N4O4
CAS Number:
Molecular Weight:
434.49
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

R$1,229.00


Estimated to ship onJune 01, 2025


Request a Bulk Order

grade

pharmaceutical primary standard

API family

tryptophan

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

[n]3(c4c(c(c3)C[C@H](N)C(=O)O)cccc4)C([n]1c2c(c(c1)C[C@H](N)C(=O)O)cccc2)C

InChI

1S/C24H26N4O4/c1-14(27-12-15(10-19(25)23(29)30)17-6-2-4-8-21(17)27)28-13-16(11-20(26)24(31)32)18-7-3-5-9-22(18)28/h2-9,12-14,19-20H,10-11,25-26H2,1H3,(H,29,30)(H,31,32)/t19-,20-/m0/s1

InChI key

DETVQFQGSVEQBH-PMACEKPBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

1,1′-Ethylidenebistryptophan EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanan Tang et al.
Journal of environmental sciences (China), 42, 259-266 (2016-04-20)
A broad range of organic compounds are known to exist in drinking water sources and serve as precursors of disinfection byproducts (DBPs). Epidemiological findings of an association of increased risk of bladder cancer with the consumption of chlorinated water has

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service