DLW354620
Corning® BioCoat™ Plate 96w PDL Wht 5pac 5cas
Synonym(s):
plastic coated cell culture plate
About This Item
shipped in
wet ice
storage temp.
room temp
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service