Skip to Content
Merck
All Photos(1)

Key Documents

CRM702

Supelco

Anions - Loamy Sand 1

certified reference material, pkg of 50 g

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
R$1,496.00

R$1,496.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
R$1,496.00

About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

R$1,496.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

CofA

current certificate can be downloaded

packaging

pkg of 50 g

manufacturer/tradename

RTC CRM702-050

technique(s)

ion chromatography: suitable

matrix

soil

application(s)

environmental
food and beverages

format

matrix material

storage temp.

2-30°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service