Skip to Content
Merck
All Photos(1)

Key Documents

CRM012

Supelco

Trace Metals - Fly Ash 2

certified reference material, pkg of 100 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

CofA

current certificate can be downloaded

packaging

pkg of 100 g

manufacturer/tradename

RTC CRM012-100

technique(s)

AAS: suitable
ICP: suitable

pH

2.86

application(s)

environmental

format

matrix material

storage temp.

2-30°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Other Notes

The following elements are not certified and are included for information only:
  • Co, V
Values of analytes vary lot to lot. Some lots may not contain all analytes.

Analyte

Description

    Aluminum
    Barium
    Cadmium
    Calcium
    Chromium
    Cobalt
    Copper
    Iron
    Lead
    Magnesium
    Manganese
    Nickel
    Potassium
    Silver
    Sodium
    Vanadium
    Zinc
    See All (17)

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Muta. 1B - Ox. Sol. 2 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1 - STOT SE 3

Target Organs

Respiratory system

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service