Skip to Content
Merck
All Photos(1)

Key Documents

CLR100

Supelco

Color 100 Pt-Co Units Calibration Standard

certified reference material

Synonym(s):

Pt-Co Units Color Calibration Standard

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
R$765.00

R$765.00


Available to ship onApril 09, 2025Details


Request a Bulk Order

Select a Size

Change View
500 ML
R$765.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

R$765.00


Available to ship onApril 09, 2025Details


Request a Bulk Order

grade

certified reference material

CofA

current certificate can be downloaded

manufacturer/tradename

RTC CLR100-100

concentration

in H2O

technique(s)

UV/Vis spectroscopy: suitable

application(s)

environmental

format

matrix material

storage temp.

2-30°C

Application

The certified reference standard is used as calibration and/or quality control standard to measure the degree of coloration of liquids as per Pt-Co color scale.

Features and Benefits

  • Ready-to-use solution
  • The sample contains water as the solvent
  • Prepared and verified in accordance with ISO 17034 and ISO/IEC 17025
  • An uncertainty value of +/- 3 associated with the solution
  • Available in 100 mL and 500 mL whole volume
  • Expiry date provided on the label and CoA
  • Traceable to SI and NIST SRMs
  • Accompanied with a detailed certificate of analysis for an effective quality management system

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service