Skip to Content
Merck
All Photos(1)

Key Documents

CCW4104

Millipore

M-YSG Broth in tubes

for microscopy, Liquid medium used for the detection and enrichment of Alicyclobacillus, in fruit juices and other acidic foods, suitable for microbiology

Synonym(s):

M-YSGB, Membrane-Yeast Starch Glucose Broth, Yeast-Starch-Glucose broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.74
Pricing and availability is not currently available.

grade

for microscopy

Quality Level

form

liquid

shelf life

9 mo. at 8‑25 °C

feature

ready-to-use

composition

dextrose, 1.0 g/L
soluble starch, 2.0 g/L
yeast extract, 2.0 g/L

packaging

box of 50 tubes

storage condition

(Tightly closed)

parameter

2 mL filling volume

tube volume

3 mL

color

yellowish

pH

3.7±0.2 (25 °C)

suitability

suitable for microscopy
Alicyclobacillus spp.

application(s)

food and beverages
microbiology

General description

Alicyclobacillus is a food-spoiling microorganism and requires an acidic environment to grow. YSG media supports the growth of all currently known species of Alicyclobacillus (A. acidocaldarius, A. acidoterrestris, A. cycloheptanicus, and A. hesperidium). The low pH value of the media, in combination with the high incubation temperature, inhibits the growth of contaminating microbiota.

Application

M-YSG Broth is used for the detection and enrichment of Alicyclobacillus, in fruit juices and other acidic foods.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service