Skip to Content
Merck
All Photos(1)

Key Documents

CAN1003

Supelco

Residual Solvents, California Set 1

Sign Into View Organizational & Contract Pricing

Select a Size

1.2 ML
R$1,332.00

R$1,332.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1.2 ML
R$1,332.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

R$1,332.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

form

liquid

packaging

ampule of 3x1.2 mL

technique(s)

gas chromatography (GC): suitable

application(s)

cannabis testing

format

multi-component solution

General description

Hydrocarbons are often used in the production of extracts in the cannabis industry. Care should be taken to minimize residual solvents remaining on material intended for human consumption. To support this, many states have enacted regulations prescribing the routine analytical testing of residual solvents in cannabis-derived products.

This multi-analyte Certified Reference Material is designed specifically to satisfy the needs of analytical testing in California.

Five components have been carefully selected for convenience of use, while promoting long-term stability in solution. The analytes are formulated at 100 μg/g each, and are designed for a 100x dilution factor relative to the California action limits.

For complete calibration of the required California residual solvents, this CRM may be used in conjunction with the other products in the series:

CAN1008
CAN1012
CAN1013

Analyte

Description

    Methylene chloride 100.00 µg/g
    Chloroform 100.00 µg/g
    Benzene 100.00 µg/g
    1,2-Dichloroethane 100.00 µg/g
    Trichloroethylene 100.00 µg/g

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service