Skip to Content
Merck
All Photos(1)

Documents

C6289

Sigma-Aldrich

Activated charcoal

suitable for plant cell culture

Synonym(s):

Charcoal activated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:
MDL number:
UNSPSC Code:
11101522
PubChem Substance ID:

vapor pressure

<0.1 mmHg ( 20 °C)

description

acid-washed with hydrochloric acid

form

powder

autoignition temp.

842 °F

resistivity

1375 μΩ-cm, 20°C (graphite)

technique(s)

cell culture | plant: suitable

mp

3550 °C (lit.)

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kohsuke Ohmatsu et al.
Nature chemistry, 6(1), 47-51 (2013-12-19)
The development of a general catalytic method for the direct and stereoselective construction of contiguous all-carbon quaternary stereocentres remains a formidable challenge in chemical synthesis. Here, we report a highly enantio- and diastereoselective [3+2] annulation reaction of 5-vinyloxazolidinones and activated
Nathalie Butt et al.
Ecological applications : a publication of the Ecological Society of America, 23(4), 936-943 (2013-07-20)
A typical way to quantify aboveground carbon in forests is to measure tree diameters and use species-specific allometric equations to estimate biomass and carbon stocks. Using "citizen scientists" to collect data that are usually time-consuming and labor-intensive can play a
Yin Wang et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(1), 533-538 (2013-12-25)
Stomatal pores surrounded by a pair of guard cells in the plant epidermis control gas exchange between plants and the atmosphere in response to light, CO2, and the plant hormone abscisic acid. Light-induced stomatal opening is mediated by at least
Nuno Carvalhais et al.
Nature, 514(7521), 213-217 (2014-09-26)
The response of the terrestrial carbon cycle to climate change is among the largest uncertainties affecting future climate change projections. The feedback between the terrestrial carbon cycle and climate is partly determined by changes in the turnover time of carbon
Michael C Dietze et al.
Annual review of plant biology, 65, 667-687 (2013-11-28)
Nonstructural carbon (NSC) provides the carbon and energy for plant growth and survival. In woody plants, fundamental questions about NSC remain unresolved: Is NSC storage an active or passive process? Do older NSC reserves remain accessible to the plant? How

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service