Skip to Content
Merck
All Photos(1)

Key Documents

C4144

Supelco

Caffeine-sodium benzoate

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
R$2,245.00

R$2,245.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
100 G
R$2,245.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

R$2,245.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

concentration

50:50 (wt:wt mixture)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

single component solution

SMILES string

[Na+].[O-]C(=O)c1ccccc1.CN2C(=O)N(C)c3ncn(C)c3C2=O

InChI

1S/C8H10N4O2.C7H6O2.Na/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2;8-7(9)6-4-2-1-3-5-6;/h4H,1-3H3;1-5H,(H,8,9);/q;;+1/p-1

InChI key

JWBPVFVNISJVEM-UHFFFAOYSA-M

Application

Caffeine-sodium benzoate may be used as a reference standard in the determination of caffeine and sodium benzoate content in caffeine and sodium benzoate injections by iso-absorption method using ultraviolet spectrophotometer.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Estimation of caffeine and sodiun benzoate in caffeine and sodium benzoate injection by isoabsorption method (isobestic method).
Sowmya V K, et al.
INTERNATIONAL JOURNAL OF PHARMACEUTICAL, CHEMICAL AND BIOLOGICAL SCIENCES, 1(1), 26-31 (2011)
K Gh M Mahmoud et al.
Iranian journal of veterinary research, 16(4), 325-330 (2016-05-14)
Cryopreservation and sexing of embryos are integrated into commercial embryo transfer technologies. To improve the effectiveness of vitrification of in vitro produced buffalo embryos, two experiments were conducted. The first evaluated the effect of exposure time (2 and 3 min)
Virginia Savy et al.
Reproduction (Cambridge, England), 159(6), 767-778 (2020-04-03)
CRISPR-mediated transcriptional activation, also known as CRISPR-on, has proven efficient for activation of individual or multiple endogenous gene expression in cultured cells from several species. However, the potential of CRISPR-on technology in preimplantation mammalian embryos remains to be explored. Here

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service