Skip to Content
Merck
All Photos(1)

Documents

BP616

Estropipate

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

3-(Sulfooxy)-estra-1,3,5(10)-trien-17-one : piperazine (1:1), Estrone sulfate piperazine salt, Harmogen, Ogen

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22O5S·C4H10N2
CAS Number:
Molecular Weight:
436.56
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

estropipate

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

C1CNCCN1.C[C@]23CC[C@H]4[C@@H](CCc5cc(OS(O)(=O)=O)ccc45)[C@@H]2CCC3=O

InChI

1S/C18H22O5S.C4H10N2/c1-18-9-8-14-13-5-3-12(23-24(20,21)22)10-11(13)2-4-15(14)16(18)6-7-17(18)19;1-2-6-4-3-5-1/h3,5,10,14-16H,2,4,6-9H2,1H3,(H,20,21,22);5-6H,1-4H2/t14-,15-,16+,18+;/m1./s1

InChI key

HZEQBCVBILBTEP-ZFINNJDLSA-N

Gene Information

human ... ESR1(2099)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Estropipate BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Biochem/physiol Actions

Estropipate is the piperazine salt of the estrogen receptor agonist estrone-3-sulfate and a potent, specific inhibitor of the organic anion transporting polypeptide 1B1 (OATP1B1, IC50 = 60 nM).

Packaging

Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

Other Notes

Sales restrictions may apply.

related product

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huifangjie Li et al.
International journal of molecular sciences, 21(17) (2020-08-28)
We found interactions between dopamine and oxidative damage in the striatum involved in advanced neurodegeneration, which probably change the microglial phenotype. We observed possible microglia dystrophy in the striatum of neurodegenerative brains. To investigate the interactions between oxidative damage and
Huifangjie Li et al.
Journal of neurochemistry, 152(2), 235-251 (2019-10-16)
The striatum with a number of dopamine containing neurons, receiving projections from the substantia nigra and ventral tegmental area; plays a critical role in neurodegenerative diseases of motor and memory function. Additionally, oxidative damage to nucleic acid may be vital

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service