Skip to Content
Merck
All Photos(1)

Documents

BP423

Phytomenadione

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

Vitamin K1, 2-Methyl-3-phytyl-1,4-naphthoquinone, 3-Phytylmenadione, Phylloquinone, Vitamin K1(20)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H46O2
CAS Number:
Molecular Weight:
450.70
Beilstein:
2568816
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

phytomenadione

form

viscous liquid (Clear)

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

refractive index

n20/D 1.527 (lit.)

mp

−20 °C (lit.)

density

0.984 g/mL at 25 °C (lit.)

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

O=C(C1=CC=CC=C21)C(C)=C(C/C=C(CCC[C@@H](CCC[C@H](C)CCCC(C)C)C)\C)C2=O

InChI

1S/C31H46O2/c1-22(2)12-9-13-23(3)14-10-15-24(4)16-11-17-25(5)20-21-27-26(6)30(32)28-18-7-8-19-29(28)31(27)33/h7-8,18-20,22-24H,9-17,21H2,1-6H3/b25-20+/t23-,24-/m1/s1

InChI key

MBWXNTAXLNYFJB-NKFFZRIASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phytomenadione is a synthetic lipid-soluble forms of vitamin K1, and works as an essential cofactor for the post-translational γ-carboxyglutamate (Gla) residues in several blood coagulation factors. It is used to treat coagulopathies caused by anticoagulant toxicosis.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Phytomenadione BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Phytomenadione Injection
  • Phytomenadione Tablets
  • Packaging

    Unit quantity: 0.25 mL. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    Storage Class Code

    10 - Combustible liquids

    WGK

    WGK 1

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Papich GM, et al. et al.
    Saunders Handbook of Veterinary Drugs, (4), 637-638 (2016)
    HPLC-APCI-MS for the determination of vitamin K1 in human plasma: method and clinical application
    Song Q, et al.
    Journal of Chromatography. B, Biomedical Sciences and Applications, 875(2), 541-545 (2008)
    Rapid HPLC assay for the assessment of vitamin K1, A, E and beta-carotene status in children (7-19 years)
    Jakob E and Elmadfa I
    Internat. J. Vit. Nutr. Res., 65(1), 31-35 (1995)

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service