Skip to Content
Merck
All Photos(1)

Documents

BCR188

Spiked milk powder (pesticides)

BCR®, certified reference material, pesticides

Synonym(s):

Spiked milk powder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

format

matrix material

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
BCR188

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Organic Pollutants: γ-HCH, p,p′-DDT, Endrin, Dieldrin, p,p′-DDE, β-HEPO, β-HCH, Hexachlorobenzene

Matrix Group: Dairy products and eggs
Sales restrictions may apply

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service