Skip to Content
Merck
All Photos(1)

Key Documents

BCR121

Wholemeal flour (vitamins)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
85151701

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

application(s)

agriculture
cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

matrix material

shipped in

wet ice

storage temp.

−20°C

Analysis Note

For more information please see:
BCR121

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Vitamins: Thiamine (Vitamin B1), Vitamin B6 (total pyridoxine), Folate (total)

Matrix Group: Cereals, Bread, Rice and Maize

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service