Skip to Content
Merck
All Photos(1)

Key Documents

BCR052

Benzo[ghi]perylene

BCR®, certified reference material

Synonym(s):

1,12-Benzoperylene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H12
CAS Number:
Molecular Weight:
276.33
Beilstein:
1913029
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Agency

BCR®

Assay

>99.7%

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

>500 °C (lit.)

mp

277-279 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

c1cc2ccc3ccc4ccc5cccc6c(c1)c2c3c4c56

InChI

1S/C22H12/c1-3-13-7-9-15-11-12-16-10-8-14-4-2-6-18-17(5-1)19(13)21(15)22(16)20(14)18/h1-12H

InChI key

GYFAGKUZYNFMBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
BCR052

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S H Cherng et al.
Toxicology and applied pharmacology, 170(1), 63-68 (2001-01-06)
Although benzo[g,h,i]perylene (BghiP) has been found to promote the carcinogenesis of benzo[a]pyrene (BaP) in animal models, not much is known about this cocarcinogenic mechanism. In this study, human hepatoma HepG2 cells cotreated with BaP and BghiP were used as a
Homdutt Sharma et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 68(1), 43-49 (2006-12-22)
The synchronous fluorescence (SF) technique has been used in the identification of polycyclic aromatic hydrocarbons (PAHs) from air particulate sample in an urban environment of Delhi, Jawaharlal Nehru University. Suspended particulate matter samples of 24h duration were collected on glass
Py and BPe solvent polarity scales: effect of temperature on pyrene and benzo[ghi]perylene fluorescence spectra.
R Waris et al.
The Analyst, 113(9), 1465-1467 (1988-09-01)
Stepan Boitsov et al.
Marine environmental research, 68(5), 236-245 (2009-08-01)
Polycyclic aromatic hydrocarbons (PAH) were measured in sediment cores from 13 locations in South-Western Barents Sea as part of a detailed study of the Norwegian seabed under the MAREANO program. The generally low PAH levels found, an average around 200
Karl L Platt et al.
Chemical research in toxicology, 18(4), 700-710 (2005-04-19)
Carcinogenic polycyclic aromatic hydrocarbons (PAH), e.g., benzo[a]pyrene (BaP), possess a bay region comprising an ortho-fused benzene ring. Benzo[ghi]perylene (BghiP) represents the group of PAHs lacking such a "classic" bay region and hence cannot be metabolically converted like BaP to bay

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service