Skip to Content
Merck
All Photos(1)

Key Documents

B4921

Sigma-Aldrich

PhastGel® Blue R

Pre-measured tablets, readily soluble.

Synonym(s):

Brilliant Blue R, Acid Blue 83, Brilliant indocyanin 6B, Coomassie Brilliant Blue R

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H44N3NaO7S2
CAS Number:
Molecular Weight:
825.97
Colour Index Number:
42660
Beilstein:
5718025
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Quality Level

SMILES string

[Na+].CCOc1ccc(Nc2ccc(cc2)C(\c3ccc(cc3)N(CC)Cc4cccc(c4)S([O-])(=O)=O)=C5\C=C/C(C=C5)=[N+](\CC)Cc6cccc(c6)S([O-])(=O)=O)cc1

InChI

1S/C45H45N3O7S2.Na/c1-4-47(31-33-9-7-11-43(29-33)56(49,50)51)40-23-15-36(16-24-40)45(35-13-19-38(20-14-35)46-39-21-27-42(28-22-39)55-6-3)37-17-25-41(26-18-37)48(5-2)32-34-10-8-12-44(30-34)57(52,53)54;/h7-30H,4-6,31-32H2,1-3H3,(H2,49,50,51,52,53,54);/q;+1/p-1

InChI key

NKLPQNGYXWVELD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Detects most proteins at 50-100 ng per band
PhastGel® Blue R has been used for the staining of protein bands after SDS-PAGE (sodium dodecyl sulfate–polyacrylamide gel electrophoresis). It has also been used for the staining of cell clones in the culture media plates.
For detection of protein bands following electrophoresis.

Reconstitution

One tablet makes 400 mL of a 0.1% staining solution.

Legal Information

PhastGel is a registered trademark of Cytiva

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Production of infectious HCV genotype 1b virus in cell culture using a novel Set of adaptive mutations.
Mori KI, et al.
BMC Microbiology, 16, 224-224 (2016)
Pharmacokinetics and Biodistribution Study of 7A7 Anti-Mouse Epidermal Growth Factor Receptor Monoclonal Antibody and Its F(ab')(2) Fragment in an Immunocompetent Mouse Model.
Rabasa Capote A, et al.
ISRN Pharmacology, 2012, doi: 10-doi: 10 (2012)
Karel Mazanec et al.
Journal of mass spectrometry : JMS, 41(12), 1570-1577 (2006-12-01)
The use of low-molecular-mass color pI markers for the determination of pI values of proteins in gel isoelectric focusing (IEF) in combination with mass spectrometry is described. Different types of substituted phenols of known pI values within the mass range
C Scheler et al.
Electrophoresis, 19(6), 918-927 (1998-06-25)
Identification of proteins separated by two-dimensional electrophoresis (2-DE) is a necessary task to overcome the purely descriptive character of 2-DE and a prerequisite to the construction of 2-DE databases in proteome projects. Matrix assisted laser desorption/ionization-mass spectrometry (MALDI-MS) has a
Grit Nebrich et al.
Electrophoresis, 28(10), 1607-1614 (2007-04-21)
The availability of easy-to-handle, sensitive, and cost-effective protein staining protocols for 2-DE, in conjunction with a high compatibility for subsequent MS analysis, is still a prerequisite for successful proteome research. In this article we describe a quick and easy-to-use methodological

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service