Skip to Content
Merck
All Photos(1)

Key Documents

99387

Supelco

Lithium ionophore IV

Selectophore, function tested

Synonym(s):

5-Butyl-5-ethyl-N,N,N′,N′-tetracyclohexyl-3,7-dioxaazelaic diamide, ETH 2137, N,N,N′,N′−Tetracyclohexyl(2-butyl-2-ethyltrimethylenedioxy)diacetamide

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
R$9,514.00

R$9,514.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
50 MG
R$9,514.00

About This Item

Empirical Formula (Hill Notation):
C37H66N2O4
CAS Number:
Molecular Weight:
602.93
Beilstein:
6455135
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

R$9,514.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

description

for ion-selective electrodes

Quality Level

product line

Selectophore

form

liquid

quality

function tested

SMILES string

CCCCC(CC)(COCC(=O)N(C1CCCCC1)C2CCCCC2)COCC(=O)N(C3CCCCC3)C4CCCCC4

InChI

1S/C37H66N2O4/c1-3-5-26-37(4-2,29-42-27-35(40)38(31-18-10-6-11-19-31)32-20-12-7-13-21-32)30-43-28-36(41)39(33-22-14-8-15-23-33)34-24-16-9-17-25-34/h31-34H,3-30H2,1-2H3

InChI key

MVPCBDWXPBIVGD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Physical form

50 mg in 0.5 mL heptane

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service