Skip to Content
Merck
All Photos(1)

Documents

95338

Supelco

2-(4-tert-Butylbenzyl)propionaldehyde

analytical standard

Synonym(s):

4-tert-Butyl-α-methyl-benzenepropanal, 4-tert-Butyl-α-methyl-hydrocinnamaldehyde, Butylphenyl methylpropional, Lilial

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20O
CAS Number:
Molecular Weight:
204.31
Beilstein:
880140
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

Assay

≥90.0% (GC)

shelf life

limited shelf life, expiry date on the label

feature

standard type calibration

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.505

density

0.946 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

[H]C(=O)C(C)Cc1ccc(cc1)C(C)(C)C

InChI

1S/C14H20O/c1-11(10-15)9-12-5-7-13(8-6-12)14(2,3)4/h5-8,10-11H,9H2,1-4H3

InChI key

SDQFDHOLCGWZPU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Jinks et al.
Brain research. Cognitive brain research, 12(2), 273-288 (2001-10-06)
Depending on the odorant, transduction during the olfactory reception process is reported to be mediated by the second messengers cyclic adenosine 3',5'-monophosphate (cAMP) or inositol 1,4,5-triphosphate (IP(3)). The present study with humans investigates the relationship between temporal processing in mixtures
Hiroko Takeuchi et al.
The Journal of general physiology, 122(3), 255-264 (2003-08-27)
It has long been believed that vertebrate olfactory signal transduction is mediated by independent multiple pathways (using cAMP and InsP3 as second messengers). However, the dual presence of parallel pathways in the olfactory receptor cell is still controversial, mainly because
E G Arnau et al.
Contact dermatitis, 43(6), 351-358 (2001-01-05)
Fragrance materials are among the most common causes of allergic contact dermatitis. The aim of this study was to identify in a perfume fragrance allergens not included in the fragrance mix, by use of bioassay-guided chemical fractionation and chemical analysis/structure-activity
Prediction of perception: probing the hOR17-4 olfactory receptor model with silicon analogues of bourgeonal and lilial.
Leszek Doszczak et al.
Angewandte Chemie (International ed. in English), 46(18), 3367-3371 (2007-04-03)
A Iida et al.
Chemical senses, 25(1), 55-59 (2000-02-11)
Using the whole-cell mode of the patch-clamp technique, we attempted to record inward currents in response to cAMP, inositol 1,4, 5-trisphosphate (IP(3)) and odorants from sensory neurons in the olfactory epithelium of the Xenopus laevis lateral diverticulum (water nose). Dialysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service