94192
10-Undecenoic acid
purum, ≥97.0% (GC)
Synonym(s):
Undecylenic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
grade
purum
Assay
≥97.0% (GC)
form
solid
refractive index
n20/D 1.449 (lit.)
bp
137 °C/2 mmHg (lit.)
mp
23-25 °C (lit.)
density
0.912 g/mL at 25 °C (lit.)
SMILES string
OC(=O)CCCCCCCCC=C
InChI
1S/C11H20O2/c1-2-3-4-5-6-7-8-9-10-11(12)13/h2H,1,3-10H2,(H,12,13)
InChI key
FRPZMMHWLSIFAZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
300.2 °F - closed cup
Flash Point(C)
149 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Antimicrobial agents and chemotherapy, 52(7), 2442-2448 (2008-05-07)
Our continuing effort in antifungal natural product discovery has led to the identification of five 6-acetylenic acids with chain lengths from C(16) to C(20): 6-hexadecynoic acid (compound 1), 6-heptadecynoic acid (compound 2), 6-octadecynoic acid (compound 3), 6-nonadecynoic acid (compound 4)
PloS one, 9(10), e110342-e110342 (2014-10-21)
Highly efficient and regioselective acylation of pharmacologically interesting gastrodin with vinyl undecylenic acid has been firstly performed through an enzymatic approach. The highest catalytic activity and regioselectivity towards the acylation of 7'-hydroxyl of gastrodin was obtained with Pseudomonas cepacia lipase.
Biotechnology progress, 30(5), 1005-1011 (2014-06-04)
Enzymatic synthesis of ascorbyl undecylenate, an unsaturated fatty acid ester of ascorbic acid, was reported with biomass-derived 2-methyltetrahydrofuran (MeTHF) as the cosolvent. Of the immobilized lipases tested, Candida antarctica lipase B (CAL-B) showed the highest activity for enzymatic synthesis of
Nature communications, 6, 6208-6208 (2015-02-12)
Nanostructured porous silicon (PSi) is emerging as a promising platform for drug delivery owing to its biocompatibility, degradability and high surface area available for drug loading. The ability to control PSi structure, size and porosity enables programming its in vivo
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service