Skip to Content
Merck
All Photos(2)

Key Documents

92395

Supelco

Ponceau SX

analytical standard

Synonym(s):

3-(2,4-Dimethyl-5-sulfophenylazo)-4-hydroxynaphthalene-1-sulfonic acid disodium salt, Food Red 1

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
R$1,351.00

R$1,351.00


Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
R$1,351.00

About This Item

Empirical Formula (Hill Notation):
C18H14N2Na2O7S2
CAS Number:
Molecular Weight:
480.42
Colour Index Number:
14700
Beilstein:
7161700
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
E Number:
E125
NACRES:
NA.24

R$1,351.00


Estimated to ship onJune 01, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

[Na+].[Na+].Cc1cc(C)c(cc1\N=N\c2cc(c3ccccc3c2O)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C18H16N2O7S2.2Na/c1-10-7-11(2)16(28(22,23)24)8-14(10)19-20-15-9-17(29(25,26)27)12-5-3-4-6-13(12)18(15)21;;/h3-9,21H,1-2H3,(H,22,23,24)(H,25,26,27);;/q;2*+1/p-2/b20-19+;;

InChI key

ONTQJDKFANPPKK-LLIZZRELSA-L

General description

Ponceau SX is classified as a synthetic food color, widely used for the maintenance and improvement of color appearance in foods.[1]

Application

Ponceau SX may be used as a reference standard for the determination of the analyte:
  • In cosmetic products by micro-matrix solid phase dispersion and liquid chromatography coupled to tandem mass spectrometry.[2]
  • In foodstuffs by combining high performance liquid chromatography (HPLC) with diode array detection (DAD) and ion-trap time-of-flight mass spectrometry (IT-TOF/MS).[1]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Identification and determination of 34 water-soluble synthetic dyes in foodstuff by high performance liquid chromatography- diode array detection- ion trap time-of-flight tandem mass spectrometry.
Li XQ
Food Chemistry, 182(1-2), 316- 326 (2015)
Determination of dyes in cosmetic products by micro-matrix solid phase dispersion and liquid chromatography coupled to tandem mass spectrometry.
Guerra E, et al.
Journal of Chromatography A, 1415, 27-37 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service