Skip to Content
Merck
All Photos(1)

Key Documents

91351

Supelco

Ophthalmic acid

≥96% (HPLC)

Synonym(s):

H-gamma-Glu-Abu-Gly-OH, L-γ-Glutamyl-(2S)-2-aminobutanoylglycine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
R$987.00

R$987.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
R$987.00

About This Item

Empirical Formula (Hill Notation):
C11H19N3O6
CAS Number:
Molecular Weight:
289.29
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.24

R$987.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥96% (HPLC)

application(s)

clinical testing

format

neat

storage temp.

−20°C

SMILES string

N([C@@H](CC)C(=O)NCC(=O)O)C(=O)CC[C@H](N)C(=O)O

InChI

1S/C11H19N3O6/c1-2-7(10(18)13-5-9(16)17)14-8(15)4-3-6(12)11(19)20/h6-7H,2-5,12H2,1H3,(H,13,18)(H,14,15)(H,16,17)(H,19,20)/t6-,7-/m0/s1

InChI key

JCMUOFQHZLPHQP-BQBZGAKWSA-N

Biochem/physiol Actions

Ophthamalic acid is an analogue of glutathione isolated from crystalline lens. Ophthalmic acid could be used as a biomarker in oxidative stress. Ophthalmic acid belongs to the class of organic compounds known as gamma-glutamyl peptides. These are oligo- and polypeptides consisting of any C-terminal alpha peptide having a gamma-glutamyl residue attached at the N alpha-position.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomoyoshi Soga et al.
The Journal of biological chemistry, 281(24), 16768-16776 (2006-04-13)
Metabolomics is an emerging tool that can be used to gain insights into cellular and physiological responses. Here we present a metabolome differential display method based on capillary electrophoresis time-of-flight mass spectrometry to profile liver metabolites following acetaminophen-induced hepatotoxicity. We

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service