Skip to Content
Merck
All Photos(1)

Documents

908908

Sigma-Aldrich

Iron(III) chloride

free-flowing, Redi-Dri, reagent grade, 97%

Synonym(s):

Ferric chloride, Iron trichloride, Molysite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeCl3
CAS Number:
Molecular Weight:
162.20
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21

grade

anhydrous
reagent grade

vapor density

5.61 (vs air)

vapor pressure

1 mmHg ( 194 °C)

product line

Redi-Dri

Assay

97%

form

powder

quality

free-flowing

mp

304 °C (lit.)

SMILES string

Cl[Fe](Cl)Cl

InChI

1S/3ClH.Fe/h3*1H;/q;;;+3/p-3

InChI key

RBTARNINKXHZNM-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Iron(III) chloride is a mild oxidizing agent and participates in various carbon-carbon-bond forming reactions. On crystallization with water, it forms hydrates. It can be prepared by reacting chlorine with the scrap iron at approximately 650°C.

Application

Redi-Dri is an innovative packaging product line that helps eliminate clumping material by adding extra packaging process steps without any chemical additives, anti-clumping agents, or hydrophobic compounds. offers a more convenient and safer way to handle the products.
Iron(III) chloride (Ferric Chloride, FeCl3) has been used as a surface modulator for the characterization of wines by fingerprinting technology. It has been used for the quantitative estimation of sulfide in various biological samples.
The vapor-phase co-reductions with other metal halides by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resitance properties.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service