Skip to Content
Merck
All Photos(2)

Key Documents

908770

Sigma-Aldrich

CX82

Umicore

Synonym(s):

Dichlorobis[(tert-butyl)dicyclohexylphosphine]palladium(II)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R$464.00

R$464.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

Select a Size

Change View
1 G
R$464.00

About This Item

Empirical Formula (Hill Notation):
C32H62P2Cl2Pd
CAS Number:
Molecular Weight:
686.11
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

R$464.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

form

powder

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

>300 °C

SMILES string

[Pd].P(C4CCCCC4)(C3CCCCC3)C(C)(C)C.P(C2CCCCC2)(C1CCCCC1)C(C)(C)C.Cl.Cl

InChI

1S/2C16H31P.2ClH.Pd/c2*1-16(2,3)17(14-10-6-4-7-11-14)15-12-8-5-9-13-15;;;/h2*14-15H,4-13H2,1-3H3;2*1H;

InChI key

UXEDXFSHGPWQJZ-UHFFFAOYSA-N

Application

Umicore CX82 is a homogeneous catalyst useful for cross coupling reactions with arenes and Suzuki-Miyaura cross coupling.

Storage and Stability

Do not store above 25°C (77°F). Store in a tightly closed original container, protected from direct sunlight in a dry, cool and well-ventilated area.

Legal Information

Product of Umicore

Additional information available at www.pmc.umicore.com

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service