Skip to Content
Merck
All Photos(1)

Key Documents

90615

Sigma-Aldrich

1,4,7-Triazacyclononane trihydrochloride

purum, ≥98.0% (AT)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6H15N3 · 3HCl
CAS Number:
Molecular Weight:
238.59
Beilstein:
5287631
MDL number:
UNSPSC Code:
12350000
PubChem Substance ID:

grade

purum

Assay

≥98.0% (AT)

mp

288 °C (dec.) (lit.)

SMILES string

Cl[H].Cl[H].Cl[H].C1CNCCNCCN1

InChI

1S/C6H15N3.3ClH/c1-2-8-5-6-9-4-3-7-1;;;/h7-9H,1-6H2;3*1H

InChI key

HNPMVNQYFPWBKI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Possible reagent for compleximetric titrations with high cation-binding selectivity.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service