Skip to Content
Merck
All Photos(1)

Key Documents

89136

Supelco

Fenamiphos-(S-methyl-d3)

PESTANAL®, analytical standard

Synonym(s):

Fenamiphos-d3, Phenamiphos-d3

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$2,880.00

R$2,880.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
R$2,880.00

About This Item

Empirical Formula (Hill Notation):
C13D3H19NO3PS
Molecular Weight:
306.38
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

R$2,880.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture

format

neat

storage temp.

2-8°C

SMILES string

O=P(NC(C)C)(OC1=CC(C)=C(SC([2H])([2H])[2H])C=C1)OCC

InChI key

ZCJPOPBZHLUFHF-VPYROQPTSA-N

General description

Fenamiphos-(S-methyl-d3) is a deuterated isotope analog of fenamiphos-(S-methyl), a derivative of the broad-spectrum, non-volatile organophosphorus nematicide fenamiphos, wherein S-methyl protons are replaced by deuterium.[1][2][3]

Application

Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The major advantage of using this technique is that the isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analogs, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bioremedial potential of fenamiphos and chlorpyrifos degrading isolates: influence of different environmental conditions
Singh BK, et al.
Soil Biology and Biochemistry, 38(9), 2682-2693 (2006)
Land use effects on sorption of pesticides and their metabolites in sandy soils. I. Fenamiphos and two metabolites, fenamiphos sulfoxide and fenamiphos sulfone, and fenarimol and azinphos methyl
Oliver DP, et al.
Australian Journal of Soil Research, 41(5), 847-860 (2003)
Tanya P Cáceres et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 43(7), 605-610 (2008-09-23)
Fenamiphos (0-ethyl-0(3-methyl-4-methylthiophenyl)-isopropylamido-phosphate) is a widely used nematicide and insecticide in bowling greens and agriculture, but information on its sorption including its metabolites is limited. Hence, the sorption of fenamiphos (nematicide) and its major degradation products fenamiphos sulfoxide (FSO) and fenamiphos

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service