Skip to Content
Merck
All Photos(1)

Key Documents

89061

Supelco

Eriodictyol

analytical standard

Synonym(s):

3·,4·,5,7-Tetrahydroxyflavanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O6
CAS Number:
Molecular Weight:
288.25
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤5.0% water

application(s)

food and beverages

format

neat

InChI

1S/C15H12O6/c16-8-4-11(19)15-12(20)6-13(21-14(15)5-8)7-1-2-9(17)10(18)3-7/h1-5,13,16-19H,6H2/t13-/m1/s1

InChI key

SBHXYTNGIZCORC-CYBMUJFWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Eriodictyol is a naturally occurring flavonoid found in citrus fruits. It can be isolated from the chinese herb, Dracocephalum rupestre. It may possess antioxidant activity.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: mentha silybum thymus
For this product we use the non-stereospecific CAS number. It is expected that the natural enantiomer is predominant, however the substance is prone to racemization in solution. The product is therefore not specified stereospecifically and is only recommended to be used for non-stereospecific analysis.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

S Ogata et al.
Bioscience, biotechnology, and biochemistry, 64(5), 1075-1078 (2000-07-06)
The flavonoid from lemon fruit (Citrus limon BURM. f.) and its metabolites, particularly eriodictyol, 3,4-dihydroxyhydrocinnamic acid, and phloroglucinol had the function of DNA fragmentation in HL-60 cells when analyzed by flow cytometry. An apoptotic DNA ladder and chromatin condensation were
The flavonoid, eriodictyol, induces long-term protection in ARPE-19 cells through its effects on Nrf2 activation and phase 2 gene expression
Johnson J, et al.
Investigative Ophthalmology & Visual Science, 50, 2398-2406 (2009)
B A 'T Hart et al.
Chemico-biological interactions, 73(2-3), 323-335 (1990-01-01)
The mechanism by which (a panel of) flanonoids inhibit the production of luminol-dependent chemiluminescence (CLlum) by activated human neutrophils is subject to this study. CLlum is frequently used as a bio-assay to quantify the effect of xenobiotics on the production
Xu Jing et al.
Toxicology and applied pharmacology, 273(3), 672-679 (2014-01-28)
Stroke is a complex disease that may involve oxidative stress-related pathways in its pathogenesis. The nuclear factor erythroid-2-related factor 2/antioxidant response element (Nrf2/ARE) pathway plays an important role in inducing phase II detoxifying enzymes and antioxidant proteins and thus has
C Huang et al.
Xenobiotica; the fate of foreign compounds in biological systems, 39(4), 312-322 (2009-04-08)
Dietary flavonoids catechin, epicatechin, eriodictyol, and hesperetin were investigated as substrates and inhibitors of human sulfotransferases (hSULTs). Purified recombinant proteins and human intestine cytosol were used as enzyme sources. hSULT1A1 and hSULT1A3 as well as human intestine cytosol can catalyse

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service