Skip to Content
Merck
All Photos(1)

Key Documents

87120

Sigma-Aldrich

Tetracyanoethylene

purum, ≥97.0% (N)

Synonym(s):

Ethylenetetracarbonitrile, NSC 24833, Percyanoethylene, TCNE

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NC)2C=C(CN)2
CAS Number:
Molecular Weight:
128.09
Beilstein:
1679885
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥97.0% (N)

mp

197-199 °C (lit.)
197-200 °C

storage temp.

2-8°C

SMILES string

N#CC(C#N)=C(C#N)C#N

InChI

1S/C6N4/c7-1-5(2-8)6(3-9)4-10

InChI key

NLDYACGHTUPAQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used for postfunctional addition to polyphenylacetylene derivatives to change the oxygen permeability

Reactant for:
  • Regioselective [2+2] cycloaddition reaction for production of BODIPY dyes and TCBD derivatives
  • Thermal addition reaction with alkynes
  • One-pot reactions with nucleophilic reagents forming aromatic cyanovinyl compounds
  • Synthesis of cobalt tetracyanoethylene films
  • Biotransformation by Botrytis cinerea

Other Notes

Powerful dienophile for cycloaddition reactions, reviews

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, 749-749 (1987)
A.J. Fatiadi
Synthesis, 249-249 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service