Skip to Content
Merck
All Photos(1)

Key Documents

87086

Supelco

Tetracontane

analytical standard

Synonym(s):

n-Tetracontane

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
R$510.00

R$510.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
R$510.00

About This Item

Linear Formula:
CH3(CH2)38CH3
CAS Number:
Molecular Weight:
563.08
Beilstein:
1800152
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

R$510.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

80-83 °C
80-84 °C (lit.)

application(s)

petroleum

format

neat

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C40H82/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37-39-40-38-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-40H2,1-2H3

InChI key

KUPLEGDPSCCPJI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Tetracontane may be used as internal standard in the quantitative determination of hydrocarbons in soil samples, using gas chromatography coupled with flame ionization detector(GC-FID).[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Manual for Soil Analysis - Monitoring and Assessing Soil Bioremediation (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service