Skip to Content
Merck
All Photos(1)

Key Documents

86897

Supelco

Tetrabutylammonium tetraphenylborate

for electrochemical analysis, ≥99.0%

Synonym(s):

Tetraphenylboron tetrabutylammonium

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
R$2,106.00

R$2,106.00


Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
5 G
R$2,106.00

About This Item

Linear Formula:
(CH3CH2CH2CH2)4N[B(C6H5)4]
CAS Number:
Molecular Weight:
561.69
Beilstein:
3857215
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

R$2,106.00


Estimated to ship onJune 01, 2025


Request a Bulk Order

Quality Level

Assay

≥99.0% (NT)
≥99.0%

form

powder

mp

230-236 °C (lit.)
233-237 °C

solubility

acetonitrile: 0.1 g/mL, clear, colorless

SMILES string

CCCC[N+](CCCC)(CCCC)CCCC.c1ccc(cc1)[B-](c2ccccc2)(c3ccccc3)c4ccccc4

InChI

1S/C24H20B.C16H36N/c1-5-13-21(14-6-1)25(22-15-7-2-8-16-22,23-17-9-3-10-18-23)24-19-11-4-12-20-24;1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4/h1-20H;5-16H2,1-4H3/q-1;+1

InChI key

ZHCCBGAUZWZGQV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Supporting electrolyte (e.g. calibration of conductometric cells)[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B.P. Artamonov et al
Zhur. Prikl. Khim., 47, 588-588 (1974)
Yuriy Zholudov et al.
The Analyst, 136(3), 598-604 (2010-11-26)
The study of electrogenerated chemiluminescence (ECL) in aqueous and non-aqueous media with tetraphenylborate anion as a co-reactant is reported. The obtained results indicate that tetraphenylborate acts according to an oxidative reduction scheme and forms a strong reducer after being homogeneously

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service