Skip to Content
Merck
All Photos(1)

Key Documents

86033

Sigma-Aldrich

Sulfamide

purum, ≥99%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
R$381.00

R$381.00


Estimated to ship onApril 15, 2025


Request a Bulk Order

Select a Size

Change View
10 G
R$381.00

About This Item

Linear Formula:
(NH2)2SO2
CAS Number:
Molecular Weight:
96.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$381.00


Estimated to ship onApril 15, 2025


Request a Bulk Order

grade

purum

Quality Level

Assay

≥99%

form

powder

mp

90-92 °C (lit.)
90-92 °C

density

1.611 g/mL at 25 °C (lit.)

SMILES string

NS(N)(=O)=O

InChI

1S/H4N2O2S/c1-5(2,3)4/h(H4,1,2,3,4)

InChI key

NVBFHJWHLNUMCV-UHFFFAOYSA-N

Gene Information

human ... CA1(759) , CA2(760)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of sulfamide derivatives in medicinal chemistry: a patent review
Reitz AB, et al.
Expert Opinion on Therapeutic Patents, 19(10), 1449-1453 (2009)
Sulfamides and sulfamide polymers directly from sulfur dioxide.
Leontiev AV, et al.
Chemical Communications (Cambridge, England), 2887-2889 (2006)
Laura Rosanò et al.
Cancer research, 74(24), 7453-7464 (2014-11-08)
The high mortality of epithelial ovarian cancer (EOC) is mainly caused by resistance to the available therapies. In EOC, the endothelin-1 (ET-1, EDN1)-endothelin A receptor (ETAR, EDNRA) signaling axis regulates the epithelial-mesenchymal transition (EMT) and a chemoresistant phenotype. However, there
Stephen T Vito et al.
Toxicology and applied pharmacology, 281(2), 185-194 (2014-12-03)
Tetramethylenedisulfotetramine (TETS) is a potent convulsant poison for which there is currently no approved antidote. The convulsant action of TETS is thought to be mediated by inhibition of type A gamma-aminobutyric acid receptor (GABAAR) function. We, therefore, investigated the effects

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service