Skip to Content
Merck
All Photos(1)

Key Documents

84692

Sigma-Aldrich

Sulfur dioxide

≥99.9%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SO2
CAS Number:
Molecular Weight:
64.06
Beilstein:
3535237
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.26 (21 °C, vs air)

vapor pressure

1779 mmHg ( 21 °C)

Assay

≥99.9%

form

gas

impurities

≤0.02% water
≤0.05% H2SO4

bp

−10 °C (lit.)

mp

−73 °C (lit.)

density

1.25 g/mL at 25 °C (lit.)

SMILES string

O=S=O

InChI

1S/O2S/c1-3-2

InChI key

RAHZWNYVWXNFOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Cylinder with net 2.4kg

Other Notes

Sales restrictions may apply

Recommended products

Recommended DIN 477 - Nr. 7

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Press. Gas Liquefied gas - Skin Corr. 1B

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Neil Cape et al.
Environment international, 29(2-3), 201-211 (2003-04-05)
The regulation of the emissions of 'traditional' primary air pollutants (fluorides, sulfur dioxide) has changed the pattern of exposure of ecological systems, with greatly reduced exposure close to sources, but with a smaller effect in some remote areas. Measurements show
Yuan-Qiang Sun et al.
Chemical communications (Cambridge, England), 49(26), 2637-2639 (2013-02-26)
A coumarin-hemicyanine dye was reported for ratiometric fluorescent detection of SO2 derivatives HSO3(-) and SO3(2-) based on a novel addition-rearrangement cascade reaction.
Yu Shang et al.
Environment international, 54, 100-111 (2013-02-26)
Health effects attributable to air pollution exposure in Chinese population have been least understood. The authors conducted a meta-analysis on 33 time-series and case-crossover studies conducted in China to assess mortality effects of short-term exposure to particulate matter with aerodynamic
Xin-Bao Wang et al.
European journal of pharmacology, 670(1), 1-6 (2011-09-20)
Sulfur dioxide is considered a toxic gas in air pollution and detrimental to many organs, however, it can be generated endogenously in the cardiovascular system in vivo. Gaseous sulfur dioxide has an endothelium-dependent vasorelaxing effect at low concentrations, but is
Xin-Bao Wang et al.
Life sciences, 98(2), 63-67 (2014-01-15)
Sulfur dioxide (SO2) is a common air pollutant and is detrimental to many organs. Its toxic effects including oxidative damage, deoxyribonucleic acid (DNA) damage and inflammation have been extensively studied. However, recent studies showed that SO2 can be generated endogenously

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service