Skip to Content
Merck
All Photos(1)

Key Documents

82631

Supelco

Purpurin

for spectrophotometric det. (of B, Pb)

Synonym(s):

1,2,4-Trihydroxyanthraquinone, Hydroxylizaric acid, Verantin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8O5
CAS Number:
Molecular Weight:
256.21
Colour Index Number:
58205
Beilstein:
1887127
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

grade

for spectrophotometric det. (of B, Pb)

Quality Level

technique(s)

UV/Vis spectroscopy: suitable
titration: suitable

mp

253-256 °C (lit.)

SMILES string

Oc1cc(O)c2C(=O)c3ccccc3C(=O)c2c1O

InChI

1S/C14H8O5/c15-8-5-9(16)14(19)11-10(8)12(17)6-3-1-2-4-7(6)13(11)18/h1-5,15-16,19H

InChI key

BBNQQADTFFCFGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Purpurin is a synthetic photosensitizer. It has tissue-penetrating property as its major absorption peak is between 650-715 nm. It is widely used as anthraquinone pigment in food colouring, textile industry, paint industry and medicinal treatments.

Application

Traditionally it has been used to dye cotton fabric materials. It may be used as reference standard in identifying red natural dyes using High performance liquid chromatography (HPLC) with UV-Vis Diode Array Detection (DAD) and electrospray mass spectrometric (ESI-MS) method.

Other Notes

Complexing agent for the spectrophotometric detm. of metals, e.g. La and Pb

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Morphological study of the combined effect of purpurin derivatives and light on transplantable rat bladder tumors.
Morgan, Alan R., et al.
Cancer Research, 47.2, 496-498 (1987)
A A Ramirez et al.
Talanta, 33(12), 1021-1025 (1986-12-01)
Mixtures of La(III) and Mg(II) form with purpurin (P; 1,2,4-trihydroxyanthraquinone) the mixed-metal complex LaMg(2)P(5), which is extracted with methyl isobutyl ketone at pH 7.5. The molar absorptivity of the complex is 6.1 x 10(4) l.mole(-1).cm(-1) at 570 nm and its
Inhibition of human cytochrome P450 1B1, 1A1 and 1A2 by antigenotoxic compounds, purpurin and alizarin.
Takahashi, Eizo, et al.
Mutation Research. Fundamental and Molecular Mechanisms of Mutagenesis, 508.1, 147-156 (2002)
Analytical Letters, 27, 1595-1595 (1994)
SERS detection of red organic dyes in Ag-agar gel.
Lofrumento, Cristiana, et al.
Journal of Raman Spectroscopy, 44.1, 47-54 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service