Skip to Content
Merck
All Photos(1)

Key Documents

81806

Supelco

Sodium 1-propanesulfonate monohydrate

suitable for ion pair chromatography, LiChropur, 98.0-102.0% (T)

Synonym(s):

1-Propanesulfonic acid sodium salt monohydrate

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
R$4,554.00

R$4,554.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
R$4,554.00

About This Item

Linear Formula:
CH3CH2CH2SO3Na · H2O
CAS Number:
Molecular Weight:
164.16
Beilstein:
3714817
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NB.21

R$4,554.00


Check Cart for Availability

Request a Bulk Order

description

anionic

Quality Level

Assay

98.0-102.0% (T)

form

powder

quality

LiChropur

mol wt

164.16 g/mol

technique(s)

ion pair chromatography: suitable

mp

250 °C (dec.) (lit.)

λ

10 % in H2O

UV absorption

λ: 210 nm Amax: 0.15
λ: 220 nm Amax: 0.06
λ: 230 nm Amax: 0.04
λ: 260 nm Amax: 0.02
λ: 500 nm Amax: 0.02

suitability

corresponds to standard for RP gradient test
corresponds to standard for filter test

SMILES string

O.[Na+].CCCS([O-])(=O)=O

InChI

1S/C3H8O3S.Na.H2O/c1-2-3-7(4,5)6;;/h2-3H2,1H3,(H,4,5,6);;1H2/q;+1;/p-1

InChI key

QBQVXXQXZXDEHE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service