Skip to Content
Merck
All Photos(1)

Key Documents

81454

Supelco

1,4-Polyisoprene

for GPC, standard, 300,000

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

for GPC
standard

CofA

exact values can be taken from the accompanying certificate of analysis

mol wt

Mn ~287000
Mp ~293000
Mw ~293000

contains

~0.1% 2,6-di-tert-butyl-4-methylphenol as stabilizer

Mw/Mn

~1.02

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service