Skip to Content
Merck
All Photos(1)

Key Documents

80317

Supelco

Acetaminophen-(ring-d4)

analytical standard

Synonym(s):

N-(4-Hydroxyphenyl-2,3,5,6-d4)-acetamide, Acetaminophen-d4, Paracetamol-(ring-d4), [2H4]-Acetaminophen, [2H4]-Acetoaminophenol, [2H4]-Paracetamol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8D4H5NO2
CAS Number:
Molecular Weight:
155.19
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

−20°C

InChI

1S/C8H9NO2/c1-6(10)9-7-2-4-8(11)5-3-7/h2-5,11H,1H3,(H,9,10)/i2D,3D,4D,5D

InChI key

RZVAJINKPMORJF-QFFDRWTDSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Akiko Toda et al.
Xenobiotica; the fate of foreign compounds in biological systems, 51(3), 316-323 (2020-11-13)
Plasma concentrations of acetaminophen, its glucuronide and sulfate conjugates, and cysteinyl acetaminophen were experimentally determined after oral administrations of 10 mg/kg in humanised-liver mice, control mice, rats, common marmosets, cynomolgus monkeys, and minipigs; the results were compared with reported human pharmacokinetic
Yuli Ekowati et al.
Chemosphere, 220, 176-184 (2018-12-26)
Alternative disinfection technologies may overcome some of the limitations of conventional treatment applied in swimming pools: chlorine-resistant pathogens (e.g. Cryptosporidium oocysts and Giardia cysts) and the formation of chlorinated disinfection byproducts. In this paper, results of full scale validation of
Xinzhu Yi et al.
Water research, 121, 46-60 (2017-05-17)
Landfill leachate could be a significant source of emerging contaminants (ECs) and antibiotic resistance genes (ARGs) into the environment. This study provides the first information on the occurrence of selected ECs and ARGs in raw leachate from 16-year old closed
Prapha Arnnok et al.
Environmental science & technology, 51(18), 10652-10662 (2017-08-17)
The continuous release of pharmaceuticals and personal care products (PPCPs) into freshwater systems impacts the health of aquatic organisms. This study evaluates the concentrations and bioaccumulation of PPCPs and the selective uptake of antidepressants in fish from the Niagara River
Michelle Schaefer et al.
Drug metabolism and disposition: the biological fate of chemicals, 44(3), 435-444 (2015-12-30)
In vitro models based on primary human hepatocytes (PHH) have been advanced for clearance (CL) prediction of metabolically stable compounds, representing state-of-the-art assay systems for drug discovery and development. Yet, limited cell availability and large interindividual variability of metabolic profiles

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service