Skip to Content
Merck
All Photos(2)

Key Documents

77374

Supelco

Petroselinic acid

analytical standard

Synonym(s):

5-Heptadecylene-1-carboxylic acid, 6-Octadecylenic acid, cis-6-Octadecenoic acid, C18:1n-12

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H34O2
CAS Number:
Molecular Weight:
282.46
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

functional group

carboxylic acid

shipped in

ambient

storage temp.

2-8°C

SMILES string

CCCCCCCCCCC\C=C/CCCCC(O)=O

InChI

1S/C18H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h12-13H,2-11,14-17H2,1H3,(H,19,20)/b13-12-

InChI key

CNVZJPUDSLNTQU-SEYXRHQNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ling Li et al.
Molecular cell, 80(3), 525-540 (2020-10-18)
Well-balanced and timed metabolism is essential for making a high-quality egg. However, the metabolic framework that supports oocyte development remains poorly understood. Here, we obtained the temporal metabolome profiles of mouse oocytes during in vivo maturation by isolating large number of
Magdalena Buszewska-Forajta et al.
Journal of pharmaceutical and biomedical analysis, 102, 331-339 (2014-12-03)
The automated mass spectral deconvolution and identification system (AMDIS) is a modern analytical tool, mostly used as a data processing method in environmental studies. The most attractive feature of that software is a fast, automatic data processing, which includes removal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service