Skip to Content
Merck
All Photos(1)

Key Documents

74023

Supelco

trans-2-Dodecenoyl-L-carnitine

analytical standard

Synonym(s):

(2R)-3-Carboxy-N,N,N-trimethyl-2-[[(2E)-1-oxo-2-dodecen-1-yl]oxy]-1-propanaminium inner salt, C12:1-Carnitine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H35NO4
CAS Number:
Molecular Weight:
341.49
UNSPSC Code:
12352200
NACRES:
NB.21

grade

analytical standard

Quality Level

Assay

≥93.0% (HPLC)

optical activity

[α]/D -15±2°, c = 0.1 in methanol

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

storage temp.

−20°C

SMILES string

[O-]C(C([H])([H])[C@](OC(/C([H])=C([H])/C([H])([H])C([H])([H])C([H])([H])C([H])([H])C([H])([H])C([H])([H])C([H])([H])C([H])([H])C([H])([H])[H])=O)([H])C([H])([H])[N+](C([H])([H])[H])(C([H])([H])[H])C([H])([H])[H])=O

General description

trans-2-Dodecenoyl-L-carnitine belongs to the class of medium-chain acylcarnitines.[1] Acyl-L-carnitines are mostly produced from L-carnitine, which plays an important role in fatty acid metabolic pathways. It acts as a carrier of long-chain acyl groups from activated fatty acids across the inner mitochondrial membrane into the mitochondrial matrix where they undergo β-oxidation to acetyl CoA to obtain usable energy via the citric acid cycle.
trans-2-Dodecenoyl-L-carnitine, belongs to the class of acylcarnitines. It serves as a specific diagnostic metabolite biomarker, which increases in very-long-chain acyl CoA dehydrogenase (VLCAD) deficiency, determined in biological samples.[2][3]

Application

trans-2-Dodecenoyl-L-carnitine may be used as an analytical reference standard for the quantification of the analyte in biological fluids of horses with atypical myopathy using gas chromatography coupled to mass spectrometry.[4] It may also be used as an analytical standard to quantify the analyte in biological samples of patients with continuous ambulatory peritoneal dialysis (CAPD) or automated peritoneal dialysis (APD) using high-performance liquid chromatography/tandem quadrupole mass spectrometry (HPLC-TQMS).[5]
trans-2-Dodecenoyl-L-carnitine may be used as an analytical reference standard for the quantification of the analyte in biological samples of horses with atypical myopathy using gas chromatography-mass spectrometry (GC−MS).[4] It may also be used as an analytical standard to quantify the analyte in biological samples of patients with continuous ambulatory peritoneal dialysis (CAPD) or automated peritoneal dialysis (APD) using high-performance liquid chromatography/tandem quadrupole mass spectrometry (HPLC-TQMS).[5]

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acylcarnitines-old actors auditioning for new roles in metabolic physiology
McCoin SC, et al.
Nature reviews. Endocrinology, 11(10), 617-617 (2015)
Acylcarnitine esters profiling of serum and follicular fluid in patients undergoing in vitro fertilization
Varnagy A, et al.
Reproductive Biology and Endocrinology, 11(1), 67-67 (2013)
Acquired multiple Acyl-CoA dehydrogenase deficiency in 10 horses with atypical myopathy
Westermann MC, et al.
Neuromuscular Disorders, 18(5), 355-364 (2008)
Current methods for determination of L-carnitine and acylcarnitine
Moder M, et al.
Monatshefte fur Chemie / Chemical Monthly, 136(8), 1279-1291 (2005)
L-Carnitine status in end-stage renal disease patients on automated peritoneal dialysis
Liberato DL , et al.
Journal of Nephrology, 27(6), 699-706 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service