Skip to Content
Merck
All Photos(1)

Key Documents

73660

Sigma-Aldrich

2-Nitrophenyl β-D-galactopyranoside

≥99.0% (HPLC)

Synonym(s):

o-Nitrophenyl β-D-galactopyranoside, ONPG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15NO8
CAS Number:
Molecular Weight:
301.25
Beilstein:
92207
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Pricing and availability is not currently available.

grade

for molecular biology

Assay

≥99.0% (HPLC)

form

powder

optical activity

[α]20/D −70±1°, c = 1% in H2O

solubility

hot water: 0.2 g/10 mL, clear, slightly yellow to deep greenish-yellow

storage temp.

2-8°C

SMILES string

C[C@@H](O)c1ccccc1
OC[C@H]1O[C@@H](Oc2ccccc2[N+]([O-])=O)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C12H15NO8/c14-5-8-9(15)10(16)11(17)12(21-8)20-7-4-2-1-3-6(7)13(18)19/h1-4,8-12,14-17H,5H2/t8-,9+,10+,11-,12-/m1/s1

InChI key

KUWPCJHYPSUOFW-YBXAARCKSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Substrate for the determination of β-galactosidases[1]

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H. Jagota et al.
Journal of Food Science, 46, 161-161 (1981)
Sung Hun Yi et al.
New biotechnology, 28(6), 806-813 (2011-09-06)
After the complete gene of a β-galactosidase from human isolate Bifidobacterium breve B24 was isolated by PCR and overexpressed in E. coli, the recombinant β-galactosidase was purified to homogeneity and characterized for the glycoside transferase (GT) and glycoside hydrolase (GH)
Mireia Baeza et al.
Analytical chemistry, 82(3), 1006-1011 (2010-01-01)
Low-temperature cofired ceramics (LTCC) technology is a versatile fabrication technique used to construct microflow systems. It permits the integration of several unitary operations (pretreatment, separation, (bio)chemical reaction, and detection stage) of an analytical process in a modular or monolithic way.
Thomas Maischberger et al.
Biotechnology journal, 5(8), 838-847 (2010-07-30)
A novel heterodimeric beta-galactosidase with a molecular mass of 105 kDa was purified from crude cell extracts of the soil isolate Lactobacillus pentosus KUB-ST10-1 using ammonium sulphate fractionation followed by hydrophobic interaction and affinity chromatography. The electrophoretically homogenous enzyme has
Kui Wang et al.
Applied microbiology and biotechnology, 88(1), 155-165 (2010-07-09)
A novel beta-galactosidase gene, zd410, was isolated by screening a soil metagenomic library. Sequence analysis revealed that zd410 encodes a protein of 672 amino acids with a predicted molecular weight of 78.6 kDa. The recombinant ZD410 was expressed and purified

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service