Skip to Content
Merck
All Photos(1)

Key Documents

73443

Supelco

Tin(II) ionophore II

Selectophore, function tested

Synonym(s):

6-(4-Nitrophenyl)-2,4-diphenyl-1,3-diazabicyclo[3.1.0]hex-3-ene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H17N3O2
CAS Number:
Molecular Weight:
355.39
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

product line

Selectophore

Quality Level

form

powder

quality

function tested

SMILES string

[O-][N+](=O)c1ccc(cc1)C2C3N2C(N=C3c4ccccc4)c5ccccc5

InChI

1S/C22H17N3O2/c26-25(27)18-13-11-16(12-14-18)20-21-19(15-7-3-1-4-8-15)23-22(24(20)21)17-9-5-2-6-10-17/h1-14,20-22H

InChI key

MMPJPLZUJANSCC-UHFFFAOYSA-N

General description

Visit our Sensor Applications portal to learn more.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Majid Arvand et al.
Analytica chimica acta, 579(1), 102-108 (2007-08-29)
A Sn2+ ion-selective electrode which was prepared with a polymeric membrane based on 6-(4-nitrophenyl)-2,4-diphenyl-3,5-diaza-bicyclo[3.1.0]hex-2-ene (NDDBH) as a ionophore. Effects of experimental parameters such as membrane composition, nature and amount of plasticizer, the amount of additive and concentration of internal solution

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service