Skip to Content
Merck
All Photos(1)

Key Documents

72843

Supelco

Hydroxytebuconazole

PESTANAL®, analytical standard

Synonym(s):

5-(4-Chlorophenyl)-2,2-dimethyl-3-(1H-1,2,4-triazol-1-ylmethyl)-1,3-pentanediol, HWG 2061

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22ClN3O2
CAS Number:
Molecular Weight:
323.82
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

General description

Hydroxytebuconazole (TEB-OH) is the main metabolite of tebuconazole (TEB), a triazole fungicide, widely used for controlling leaf spot diseases. It is reportedly used as an exposure biomarker in the detection of tebuconazole.

Application

Hydroxytebuconazole may be used as an analytical reference standard for the quantification of the analyte in urine samples using chromatography techniques.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toxicokinetics of a urinary metabolite of tebuconazole following controlled oral and dermal administration in human volunteers
Oerlemans A, et al.
Archives of Toxicology, 93(9), 2545-2553 (2019)
Urine collection methods for non-toilet-trained children in biological monitoring studies: Validation of a disposable diaper for characterization of tebuconazole exposure
Oerlemans A, et al.
Toxicology Letters, 298(9), 201-206 (2018)
Biological monitoring of exposure to tebuconazole in winegrowers
Fustinoni S, et al.
Journal of Exposure Science & Environmental Epidemiology, 24(6), 643-649 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service