Skip to Content
Merck
All Photos(2)

Key Documents

71038ATMBC

Millipore

GDS Cronobacter Tq II Automation Kit

BioControl Systems, for use with GDS Rotor-Gene®

Sign Into View Organizational & Contract Pricing

Select a Size

576 TESTS
R$48,076.00

R$48,076.00


Check Cart for Availability

Request more information

Select a Size

Change View
576 TESTS
R$48,076.00

About This Item

UNSPSC Code:
41106302
NACRES:
NA.85

R$48,076.00


Check Cart for Availability

Request more information

Agency

AOAC® (OMA)
AOAC® (PTM)
MicroVal

Quality Level

product line

Assurance GDS®

shelf life

6-12 mo.

packaging

pkg of 576 tests

manufacturer/tradename

BioControl Systems

application(s)

food and beverages

compatibility

for use with GDS Rotor-Gene®

storage temp.

2-8°C

suitability

Cronobacter spp.

General description

Assurance GDS® is an advanced genetic detection system for food pathogens. GDS Cronobacter Tq II is an automated nucleic acid amplification system for the detection of Cronobacter spp. The proprietary PickPen® IMS-based sample preparation procedure quickly and effectively captures Cronobacter spp., separating the pathogen from inhibitory background materials ensuring valid PCR reactions for all sample types, including challenging food and environmental samples. Assurance GDS® PCR technology provides two additional levels of specificity through the use of both highly specific primers and probes to ensure accurate detection of Cronobacter spp.

Application

Assurance GDS® Cronobacter Tq II is designed for the detection of Cronobacter spp. in infant nutritional formula, ingredients, and environmental samples.

Features and Benefits

  • Simple 1-step enrichment procedure for powdered infant formula, ingredients, and environmental samples
  • Results are available in 22-26 hours, including enrichment, sample prep and analysis

Components

Each 576ATM (for PPMX Automation) kit includes:
  • Cronobacter Amplification Tubes Tq (0.2 mL)
  • Cronobacter Concentration Reagent
  • For each 576ATM kit, the following are also necessary and should be ordered separately:
  • 61031BC Wash Solution Kit
  • 34724BC Resuspension Buffer Tq
  • Testing Method

    PCR, Immunomagnetic Separation (IMS)

    Legal Information

    AOAC is a registered trademark of AOAC International
    Assurance GDS is a registered trademark of BioControl Systems, Inc.
    PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany
    Rotor-Gene is a registered trademark of Qiagen GmbH

    Storage Class Code

    10 - Combustible liquids


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Questions

    Reviews

    No rating value

    Active Filters

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service